EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Radical promoted Wagner-Meerwein-type rearrangement of epoxides in camphoric systems using a Ti(III) radical source.
PMID 16277363
Abstract
[Chemical reaction: see text] A radical based Wagner-Meerwein-type rearrangement has been observed in camphoric systems. The radical was generated from the epoxide using Cp2TiCl as the radical source. The radical initiator Cp2TiCl was prepared in situ from commercially available Cp2TiCl2 and Zn dust in THF under argon.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
234826
Bis(cyclopentadienyl)titanium(IV) dichloride, 97%
C10H10Cl2Ti