EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Brønsted acid-catalyzed imine amidation.
PMID 16277499
Abstract
A new method for the Brønsted acid-catalyzed addition of amide nucleophiles to imines to produce protected aminal products is described. Simple Brønsted acids (phenyl phosphinic acid and trifluoromethanesulfonimide) were shown to be excellent catalysts, providing high yields of the aminal product. A catalytic asymmetric imine amidation using sulfonamides as nucleophiles was successful when a hindered biaryl phosphoric acid catalyst derived from 2,2'-diphenyl-[3,3'-biphenanthrene]-4,4'-diol (VAPOL) was used. Excellent yields and enantioselectivities were found in these additions (up to 99% ee).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
675512
(R)-(–)-VAPOL hydrogenphosphate
C40H25O4P