EMAIL THIS PAGE TO A FRIEND
The Journal of pharmacology and experimental therapeutics
Comparative evaluation of HERG currents and QT intervals following challenge with suspected torsadogenic and nontorsadogenic drugs.
PMID 16278312
Abstract
The purpose of the present study was to comparatively evaluate human HERG currents and QT intervals following challenge with suspected torsadogenic and nontorsadogenic drugs. Various concentrations of 14 different drugs were initially evaluated in terms of their relative potency to block I(HERG) in stably transfected human embryonic kidney cells. Four general categories of drugs were identified: high-potency blockers (IC50 < 0.1 microM) included lidoflazine, terfenadine, and haloperidol; moderate-potency blockers (0.1 microM < IC50 < 1 microM) included sertindole, thioridazine, and prenylamine; low-potency blockers (IC50 > 1 microM) included propafenone, loratadine, pyrilamine, lovastatin, and chlorpheniramine; and ineffective blockers (IC50 > 300 microM) included cimetidine, pentamidine, and arsenic trioxide. All measurements were performed using similar conditions and tested acute drug effects only (<30 min of drug exposure per measurement). Since two of the drugs that were ineffective I(HERG) blockers, arsenic trioxide and pentamidine, have been associated with cardiac repolarization delays (QT interval lengthening) and torsades de pointes ventricular arrhythmias in patients, we chose to evaluate them further using the isolated perfused rabbit heart model. Neither arsenic trioxide nor pentamidine had any significant effect on QT intervals in this model, even at relatively high (micromolar) concentrations. Similar results were obtained for loratadine in this model. When the hearts were challenged with a known torsadogenic drug such as cisapride, significant QT lengthening was rapidly induced. These results demonstrate that arsenic trioxide and pentamidine are essentially devoid of direct acute effects on cardiac repolarization or inhibition of I(HERG).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
LO2219 Cardiotoxicity Ligand Set
L9668
Lidoflazine, ≥98% (HPLC), powder
C30H35F2N3O