EMAIL THIS PAGE TO A FRIEND
Ceskoslovenska gynekologie
[Monitoring the effects of oxyphilline Spofa in the umbilical-placental circulation].
PMID 1628328
Abstract
Vasodilatation treatment is an inevitable part of comprehensive therapy of intrauterine growth retardation of the foetus. The authors present the results of investigations of the effect of Oxyphylline on the umbilical and placental circulation of pregnant women with confirmed disorders of the flow parameters. Oxyphylline Spofa, 3 x 100 mg orally, is the drug of choice in treatment of foetal hypotrophy with a marked effect on the umbilical and placental circulation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H9006
7-(β-Hydroxyethyl)theophylline, solid
C9H12N4O3