EMAIL THIS PAGE TO A FRIEND
Clinical and experimental immunology
Comparison of the properties of the CsA analogs monoacetyl CyC (o-acetyl-threonine2 cyclosporin) and methyl-alanyl CsA (N-methyl-L-alanyl6 cyclosporin); monoacetyl cyclosporin is immunosuppressive without binding to cyclophilin.
PMID 1628422
Abstract
Cyclosporin (CsA) is an immunosuppressant which binds to cyclophilin (Cyp). The relationship between Cyp binding and immunosuppression has been questioned since one of the analogs of CsA, N-methyl-L-alanyl6 cyclosporin (methyl-alanyl CsA) binds to Cyp but is not immunosuppressive. We compared the immunosuppressive properties of CsA, methyl-alanyl CsA and o-acetyl-threonine2 cyclosporin (monoacetyl CyC), since monoacetyl CyC does not bind to Cyp when tested in cell-free assays and its immunosuppressive properties had not been tested. Cyp is a peptidyl-prolyl isomerase which is abundant in all human tissues, yet the activities of CsA are mostly confined to inhibition of T cell and thymocyte activation, and to neuro- and nephro-toxicity and are independent of inhibition of the isomerase. Activation of thymocytes and of T cells is regulated by the binding of a nuclear factor(s) (NFs) to the NF-AT region (-285 to -255) of the IL-2 promoter. We studied inhibition of binding to the NF-AT region of NFs derived from primary cultures of thymocytes treated with CsA or its analogs. In addition, we compared the effect of CsA and its analogs on the expression of the IL-2 gene in a stably transfected Jurkat-cell line (Fgl 5) which contains three copies of NF-AT and the reporter enzyme beta-galactosidase; and on inhibition of proliferation induced by concanavalin A (Con A) or IL-2. We found that monoacetyl CyC which does not bind to Cyp is immunosuppressive by our criteria when tested in cultured cells due to either a different mechanism of action or to metabolic activation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0192 Cyclosporin C, ≥95% (HPLC), from Acremonium luzulae
C62H111N11O13