EMAIL THIS PAGE TO A FRIEND
Cancer Research
Cancer-specific functions of SIRT1 enable human epithelial cancer cell growth and survival.
PMID 16288037
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
MAB1501
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®