EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Piperazinyl oxime ethers as NK-1 receptor antagonists.
PMID 16289817
Abstract
The synthesis and structure-activity relations for a new class of centrally active NK-1 receptor antagonists are described. The new compounds are based on piperazine 2 and contain an oxime ether functionality. Several new compounds have high affinity for the NK-1 receptor and show good antagonistic activity in the gerbil foot-tapping assay.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G114
GR 73632, >97%
C40H59N7O6S