EMAIL THIS PAGE TO A FRIEND
Journal of colloid and interface science
Condensation of nucleobases at mercury/aqueous solution interface--a structural perspective using hydrogen bonding considerations.
PMID 16290652
Abstract
The two-dimensional condensation behavior exhibited by nucleobases at a mercury/aqueous solution interface is analyzed on the basis of their hydrogen-bonded quadruplex structures, and the experimentally observed critical temperatures are rationalized incorporating different types of hydrogen bonding, surface coordination effects, and stacking considerations. The proposed methodology provides a structural basis for the interpretation of critical temperatures and enables the calculation of the same pertaining to different modified nucleobases. The applicability of the procedure to order-disorder transitions of water dipoles at Hg electrodes is also indicated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I6875
5-Iodocytosine
C4H4IN3O