EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Stereospecific addition of H-P bond to alkenes: a simple method for the preparation of (R(P))-phenylphosphinates.
PMID 16292852
Abstract
[reaction: see text] A variety of functionalized optically pure (R(P))-alkylphenylphosphinates are readily prepared by stereospecific radical or base-catalyzed additions of the easily available (R(P))-menthyl phenylphosphinate to alkenes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P28808
Phenylphosphinic acid, 99%
C6H7O2P