EMAIL THIS PAGE TO A FRIEND
Prostaglandins, leukotrienes, and essential fatty acids
Effect of prostaglandins against alloxan-induced diabetes mellitus.
PMID 16297610
Abstract
Previously, we observed that alloxan-induced in vitro cytotoxicity and apoptosis in an insulin secreting rat insulinoma, RIN, cells was prevented by prior exposure to prostaglandin (PG) E(1), PGE(2), PGI(2), PGF(1)(alpha), and PGF(3)(alpha) (P<0.05 compared to alloxan), whereas thromboxane B(2) (TXB(2)) and 6-keto-PGF(1)(alpha) were ineffective. In an extension of these studies, we now report that prior intraperitoneal administration of PGE(1), PGE(2), PGF(1)(alpha), and PGF(3)(alpha) prevented alloxan-induced diabetes mellitus in male Wistar rats, whereas PGI(2), TXB(2), and 6-keto PGF(1)(alpha) were not that effective. PGE(1), PGE(2), PGF(1)(alpha), and PGF(3)(alpha) not only attenuated chemical-induced diabetes mellitus but also restored the antioxidant status to normal range in red blood cells and pancreas. These results suggest that PGE(1), PGE(2), PGF(1)(alpha), and PGF(3)(alpha) can abrogate chemically induced diabetes mellitus in experimental animals and attenuate the oxidant stress that occurs in diabetes mellitus.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P5765
Prostaglandin F, synthetic
C20H36O5