EMAIL THIS PAGE TO A FRIEND
Bulletin of experimental biology and medicine
Cooperative type of platelet hypersensitivity to ADP.
PMID 16307036
Abstract
We found that gestosis is associated with platelet hypersensitivity to ADP. Cell P2X1 receptors exhibited a positive cooperative response to ADP (EC(50)=10.88+/-3.70 nM, Hill constant n=2.59+/-0.50 rel. units). Cooperative binding of ADP to platelet P2X1 receptors was also observed during incubation of cells from pregnant women with isosorbide dinitrate.