EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Equilibrium, kinetic and solvent effect studies on the reactions of [RuIII(Hedta)(H2O)] with thiols.
PMID 16311647
Abstract
The interaction of [Ru(III)(edta)(H(2)O)](-) with a series of selected thiols having extra functional groups was investigated potentiometrically and kinetically. The pK(a) values of the uncoordinated carboxylic acid group and coordinated water molecule are 3.12 and 7.41, respectively, in aqueous solution at 25 degrees C and 0.1 M ionic strength. The formation constants of the complexes were determined in the pH range 3-9, and the concentration distribution of the various complex species was evaluated as a function of pH. The effect of dioxane on the pK(a) values of [Ru(III)(Hedta)(H(2)O)] and the formation constants of the corresponding thiol complexes is presented. The study also provides mechanistic information on the reaction of [Ru(III)(edta)(H(2)O)](-) with the thiols. The low values of DeltaH(not equal) and negative values of DeltaS(not equal) and DeltaV(not equal) for the substitution reactions of [Ru(III)(edta)(H(2)O)](-) clearly support the associative character of the substitution process.