EMAIL THIS PAGE TO A FRIEND
Biopolymers
Monensin A methyl ester complexes with Li+, Na+, and K+ cations studied by ESI-MS, 1H- and 13C-NMR, FTIR, as well as PM5 semiempirical method.
PMID 16312019
Abstract
Monensin A methyl ester (MON1) was synthesized by a new method and its ability to form complexes with Li+, Na+, and K+ cations was studied by electrospray ionization-mass spectroscopy (ESI-MS), 1H and 13C nuclear magnetic resonance (NMR), Fourier transform infrared (FTIR), and PM5 semiempirical methods. It is shown that MON1 with monovalent metal cations forms stable complexes of 1:1 stoichiometry. The structures of the complexes are stabilized by intramolecular hydrogen bonds in which the OH groups are always involved. In the structure of MON1, the oxygen atom of the C=O ester group is involved in very weak bifurcated intramolecular hydrogen bonds with two hydroxyl groups, whereas in the complexes of MON1 with monovalent metal cations the C=O ester group is not engaged in any intramolecular hydrogen bonds. Furthermore, it is demonstrated that the strongest intramolecular hydrogen bonds are formed within the MON1-Li+ complex structure. The structures of the MON1 and its complexes with Li+, Na+, and K+ cations are visualized and discussed in detail.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
30552
Monensin methyl ester, Selectophore, ≥97.0% (TLC)
C37H64O11