EMAIL THIS PAGE TO A FRIEND
Journal of chemotherapy (Florence, Italy)
Improved susceptibilities to cinoxacin and other antimicrobial agents of some Providencia species re-identified on the basis of fermentation of polyhydric alcohols.
PMID 16312291
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C8645
Cinoxacin
C12H10N2O5