EMAIL THIS PAGE TO A FRIEND
Journal of chemotherapy (Florence, Italy)
Activity in vitro of urine samples from patients treated by nitroxoline against mycoplasmas.
PMID 16312380
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
140325
8-Hydroxy-5-nitroquinoline, 96%
C9H6N2O3