EMAIL THIS PAGE TO A FRIEND
Journal of chemotherapy (Florence, Italy)
Comparison of two bacampicillin therapeutic regimens in infections of middle ear, pharynx and paranasal sinuses.
PMID 16312621
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B0070000 Bacampicillin hydrochloride, European Pharmacopoeia (EP) Reference Standard
C21H27N3O7S · HCl