EMAIL THIS PAGE TO A FRIEND
Journal of chemotherapy (Florence, Italy)
The use of enoxacin in the treatment of urinary disease: a critical review of the Italian and international literature.
PMID 16312668