EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
High-performance liquid chromatographic enantioseparation of bicalutamide and its related compounds.
PMID 16314163
Abstract
Direct high-performance liquid chromatographic methods were developed for the enantioseparation of (R,S)-bicalutamide (1) and its analogs (+/-)-3-chloro-N-(4-cyano-3-(trifluoromethyl)phenyl)-2-hydroxy-2-methylpropanamide (2), (+/-)-N-(4-cyano-3-(trifluoro-methyl)phenyl)-2-methyloxirane-2-carboxamide (3), (+/-)-4-fluorophenylsulfonyl-2-hydroxy-2-methylpropionic acid (4) and (+/-)-3-hydroxy-N-(4-cyano-3-(trifluoromethyl)phenyl)-2-hydroxy-2-methylpropanamide (5). The methods involved the use of a cellulose-based Chiralcel OD-H, macrocyclic glycopeptide-based Chirobiotic T, TAG and R, beta-cyclodextrin-based Cyclobond I 2000SN and t-butyl carbamate-derivatized quinine-based columns. The conditions affording the best resolution were found by selection and variation of the mobile-phase compositions, and the differences in separation capability of the methods were noted. The sequence of elution of the enantiomers was determined in all cases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
10300AST Astec® CHIROBIOTIC® HPLC Column Screening Kit, 5 μm particle size, L × I.D. 10 cm × 4.6 mm
10305AST Astec® CHIROBIOTIC® HPLC Column Screening Kit, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
13019AST Astec® CHIROBIOTIC® R Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
13020AST Astec® CHIROBIOTIC® R Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 2.1 mm
13022AST Astec® CHIROBIOTIC® R Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 4.6 mm
13023AST Astec® CHIROBIOTIC® R Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
13024AST Astec® CHIROBIOTIC® R Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
12018AST Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
12019AST Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
12020AST Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 2.1 mm
12010AST Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 3 mm
12021AST Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 5 cm × 4.6 mm
12022AST Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 4.6 mm
12023AST Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
12024AST Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
12034AST Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 10 mm
14018AST Astec® CHIROBIOTIC® TAG Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
14019AST Astec® CHIROBIOTIC® TAG Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
14020AST Astec® CHIROBIOTIC® TAG Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 2.1 mm
14022AST Astec® CHIROBIOTIC® TAG Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 4.6 mm
14023AST Astec® CHIROBIOTIC® TAG Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
14024AST Astec® CHIROBIOTIC® TAG Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
14232AST Astec® CHIROBIOTIC® TAG Chiral HPLC Column, 16 μm particle size, L × I.D. 15 cm × 10 mm
14034AST Astec® CHIROBIOTIC® TAG Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 10 mm
14044AST Astec® CHIROBIOTIC® TAG Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 21.2 mm
11018AST Astec® CHIROBIOTIC® V Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
11019AST Astec® CHIROBIOTIC® V Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
11020AST Astec® CHIROBIOTIC® V Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 2.1 mm
11022AST Astec® CHIROBIOTIC® V Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 4.6 mm
11023AST Astec® CHIROBIOTIC® V Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm