EMAIL THIS PAGE TO A FRIEND
Organic letters
Formal syntheses of (+/-)-pinnaic acid and (+/-)-halichlorine.
PMID 16321034
Abstract
[chemical reaction: see text]. Concise formal syntheses of marine alkaloids (+/-)-pinnaic acid (1) and (+/-)-halichlorine (2) have been accomplished from a common intermediate. The syntheses illustrate the utility of selective olefin cross metathesis methodologies for the elaboration of advanced synthetic intermediates in complex molecule synthesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
422436
Methyl 1-cyclopentene-1-carboxylate, 97%
C7H10O2