EMAIL THIS PAGE TO A FRIEND
Cancer research
Altered localization of p120 catenin during epithelial to mesenchymal transition of colon carcinoma is prognostic for aggressive disease.
PMID 16322241
Abstract
We examined the expression and localization of p120 catenin (p120ctn) as a consequence of the epithelial to mesenchymal transition (EMT) of highly differentiated colon carcinoma cells (LIM1863 cells). This unique line grows in suspension as spheroids and undergoes an EMT within 24 hours following stimulation with transforming growth factor-beta and tumor necrosis factor-alpha. Although p120ctn expression remains stable during the EMT, its localization shifts from cell-cell junctions to the cytoplasm. Interestingly, a marked decrease in RhoA activation coincident with E-cadherin loss occurs during the EMT and correlates with the formation of a p120ctn/RhoA complex. Use of RNA interference showed that p120ctn reduction results in increased RhoA activity and a significant decrease in the motility of post-EMT cells. To determine the relevance of these findings to colorectal cancer progression, we assessed p120ctn expression by immunohistochemistry in 557 primary tumors. Of note, we observed that 53% of tumors presented cytoplasmic staining for p120ctn, and statistical analysis revealed that this localization is predictive of poor patient outcome. Cytoplasmic p120ctn correlated with later-stage tumors, significantly reduced 5- and 10-year survival times and a greater propensity for metastasis to lymph nodes compared with junctional p120ctn. We also confirmed that altered localization of p120ctn corresponded with loss or cytoplasmic localization of E-cadherin. These alterations in E-cadherin are also associated with a significant reduction in patient survival time and an increase in tumor stage and lymph node metastasis. These data provide a compelling argument for the importance of both p120ctn and the EMT itself in the progression of colorectal carcinoma.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
420M-1
p120 Catenin (MRQ-5) Mouse Monoclonal Antibody