EMAIL THIS PAGE TO A FRIEND
Inorganic chemistry
Synthesis and in solution and solid state structural study of intramolecular Pt...H interactions in pentafluorophenyl platinum(II) complexes containing the ligands triazene, formamidine, and 2-aminopyridine.
PMID 16323931
Abstract
The preparation of the [NBu4][Pt(C6F5)3L] complexes (L=triazene, formamidine, 2-aminopyridine,) have been carried out. These ligands contain a hydrogen atom, with more or less acidic character, in a position suitable for establishing an intramolecular hydrogen bonding interaction with the metal center. This interaction has been detected in solution for; its 1H NMR spectrum shows that the resonance assignable to this hydrogen has platinum satellites. For, this coupling is not observed, and the interaction, if it exists, has to be weaker because of the less acidic character of the hydrogen atom. The 2-aminopyridine ligand is more flexible than the triazene or formamidine, and also in this case, no evidence of the interaction in solution is obtained. Nevertheless, if another potential proton acceptor is present, such as ClO4- in [NBu4]2[Pt(C6F5)3(C5H6N2)](ClO4), a conventional N-H...O-Cl hydrogen bond is formed. The crystal structures of complexes have been determined by X-ray diffraction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F15803
Formamidine acetate salt, 99%
CH4N2 · C2H4O2
268607
Formamidine hydrochloride, 97%
CH4N2 · HCl