EMAIL THIS PAGE TO A FRIEND
European journal of pharmacology
Chronic corticosterone treatment increases the endocannabinoid 2-arachidonylglycerol in the rat amygdala.
PMID 16324692
Abstract
This research was designed to examine the effect of three weeks of administration of corticosterone (20 mg/kg) on endocannabinoid content and cannabinoid CB1 receptor binding in the amygdala. It was found that the endocannabinoid 2-arachidonylglycerol was significantly increased in the amygdala following chronic corticosterone treatment. However, there was no change in either the maximal binding (Bmax) or binding affinity (KD) of [3H]-CP 55,940 to the CB1 receptor in the amygdala. Given the role of amygdalar endocannabinoids in the regulation of emotionality, this suggests that the ability of glucocorticoids to influence affective behavior may involve interactions with regulation of endocannabinoid content.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46149
Corticosterone 21-acetate, VETRANAL, analytical standard
C23H32O5