EMAIL THIS PAGE TO A FRIEND
Toxicology letters
Estrogenic effects in vitro and in vivo of the fungicide fenarimol.
PMID 16324804
Abstract
The fungicide fenarimol has the potential to induce endocrine disrupting effects via several mechanisms since it possesses both estrogenic and antiandrogenic activity and inhibits aromatase activity in cell culture studies. Hence, the integrated response of fenarimol in vivo is not easy to predict. In this study, we demonstrate that fenarimol is also estrogenic in vivo, causing significantly increased uterine weight in ovariectomized female rats. In addition, mRNA levels of the estrogen responsive gene lactoferrin (LF) were decreased in uteri, serum FSH levels were increased, and T3 levels decreased in fenarimol-treated animals. To our knowledge, only two other pesticides (o,p-DDT and methoxychlor) have previously been reported to induce an estrogenic response in the rodent uterotrophic bioassay. A pronounced xenoestrogenicity in serum samples from rats treated with fenarimol and estradiol benzoate (E2B) separately or in combination was observed, demonstrating the usefulness of this approach for estimating the integrated internal exposure to xenoestrogens. The MCF-7 cell proliferation assay was used to investigate further the dose-response curves for the estrogenic, antiestrogenic, and aromatase inhibiting properties of fenarimol in vitro. The results indicates that fenarimol exhibits a dual effect being aromatase inhibitor at low concentrations and estrogenic at higher concentrations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45484
Fenarimol, PESTANAL®, analytical standard
C17H12Cl2N2O