EMAIL THIS PAGE TO A FRIEND
Plant Cell
Ordered histone modifications are associated with transcriptional poising and activation of the phaseolin promoter.
PMID 16326929
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
07-353
Anti-acetyl-Histone H3 (Lys14) Antibody, Anti-acetyl-Histone H3 (Lys14) Antibody is a rabbit polyclonal antibody for detection of acetyl-Histone H3 (Lys14) also known as H3K14Ac, Histone H3 (acetyl K14) & has been validated in ChIP, WB, ChIP-seq, DB, Mplex.
07-352
Anti-acetyl-Histone H3 (Lys9) Antibody, Anti-acetyl-Histone H3 (Lys9) Antibody is a Rabbit Polyclonal Antibody for detection of acetyl-Histone H3 (Lys9) also known as H3K9Ac, Histone H3 (acetyl K9) and has been published and validated in ChIP, WB, Mplex.
06-599
Anti-acetyl-Histone H3 Antibody, Detect acetyl-Histone H3 with Anti-acetyl-Histone H3 Antibody (Rabbit Polyclonal Antibody), that has been shown to work in WB, ICC, ChIP, ChIP-seq.
07-030
Anti-dimethyl-Histone H3 (Lys4) Antibody, Use Anti-dimethyl-Histone H3 (Lys4) Antibody (rabbit polyclonal antibody) is published and validated in ChIP, DB, ICC, IF, WB, PIA, ChIP-seq. to detect dimethyl-Histone H3 (Lys4) also known as H3K4me2, Histone H3 (di methyl K4).
07-441
Anti-dimethyl-Histone H3 (Lys9) Antibody, Use Anti-dimethyl-Histone H3 (Lys9) Antibody (Rabbit Polyclonal Antibody) validated in DB, ICC, PIA, WB to detect dimethyl-Histone H3 (Lys9) also known as H3K9me2, Histone H3 (di methyl K9).
07-367 Anti-dimethyl-Histone H4 (Lys20) Antibody
06-755
Anti-Histone H3 Antibody, Anti-Histone H3 Antibody is a Rabbit Polyclonal Antibody for detection of Histone H3 also known as histone 3-H3 & has been validated in ChIP, ICC & WB.
06-946
Anti-hyperacetylated Histone H4 (Penta) Antibody, Use Anti-hyperacetylated Histone H4 (Penta) Antibody (Rabbit Polyclonal Antibody) validated in ChIP, ICC, WB to detect hyperacetylated Histone H4 (Penta) also known as H4 histone family member A.
07-473
Anti-trimethyl-Histone H3 (Lys4) Antibody, Anti-trimethyl-Histone H3 (Lys4) Antibody is a rabbit polyclonal antibody for detection of Histone H3 trimethylated at lysine 4. Also known as Anti-H3K4me3, this highly specific and well published antibody has been validated in ChIP, DB, WB, PIA, ChIP-seq.