EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Synthesis and anti-inflammatory activity of resveratrol analogs.
PMID 16327196
Abstract
Seventeen novel resveratrol derivatives were synthesized. Their anti-inflammatory activities were tested on xylene-induced mouse ear edema. The pharmacological results showed that some compounds have potent anti-inflammatory activities.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
330264
3,5-Dimethoxybenzyl chloride, 99%
C9H11ClO2