EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Biological evaluation of isothiazoloquinolones containing aromatic heterocycles at the 7-position: In vitro activity of a series of potent antibacterial agents that are effective against methicillin-resistant Staphylococcus aureus.
PMID 16337789
Abstract
We synthesized a diverse series of 9H-isothiazolo[5,4-b]quinoline-3,4-diones containing heteroaromatic groups at the 7-position via palladium-catalyzed cross-coupling. Many of these compounds demonstrated potent antistaphylococcal activity (MICs 2 microg/mL) against a multi-drug-resistant strain (ATCC 700699) and low cytotoxic activity (CC(50)>100 microM) against the human cell line Hep2 (laryngeal carcinoma).