EMAIL THIS PAGE TO A FRIEND
Yao xue xue bao = Acta pharmaceutica Sinica
[Expression of human phosphodiesterase 3A gene using baculovirus expression system in insect cell].
PMID 16342682
Abstract
To investigate the expression of recombinant human phosphodiesterase 3A (HPDE3A) using baculovirus expression system in Tn cell line. The HPDE3A cDNA was recombined with baculovirus, and then the recombinant was transfected into Tn cell line. The expression of HPDE3A in Tn cell line was detected and identified by the RT-PCR, SDS-PAGE and Western blotting. The recombinant HPDE3A protein was stably expressed in Tn cell line and detected by the distinct morphological changes of Tn cell, RT-PCR, SDS-PAGE and Western blotting using polyclonal antibody. The M(w) of the recombinant protein was about 120 kD. Recombinant HPDE3A can be expressed in Tn cell line using the baculovirus expression system, and thus provided the basic material for studying its bioactivity and application in screening for HPDE3A inhibitor.