EMAIL THIS PAGE TO A FRIEND
Life sciences
Involvement of adenosine A1 and A2A receptors in (-)-linalool-induced antinociception.
PMID 16343551
Abstract
In recent studies performed in our laboratory we have shown that acute administration of (-)-linalool, the natural occurring enantiomer in essential oils, possesses anti-inflammatory, antihyperalgesic and antinociceptive effects in different animal models. The antihyperalgesic and antinociceptive effects of (-)-linalool have been ascribed to its capacity in stimulating the opioidergic, cholinergic and dopaminergic systems, as well as to its interaction with K+ channels, or to its local anaesthetic activity and/or to the negative modulation of glutamate transmission. Activation of A1 or A2A receptors has been shown to induce antinociceptive effects, and the possible involvement of adenosine in (-)-linalool antinociceptive effect, has not been elucidated yet. Therefore, in the present study, we have investigated the effects of 1,3-dipropyl-8-cyclopentylxanthine (DPCPX), a selective adenosine A1 receptor antagonist and the effects of 3,7-dimethyl-1-propargilxanthine (DMPX), a selective adenosine A2A receptor antagonist on the antinociception of (-)-linalool in mice, measured in the hot-plate test. Both DPCPX (0.1 mg/kg; i.p.) and DMPX (0.1 mg/kg; i.p.) pre-treatment significantly depressed the antinociceptive effect of (-)-linalool at the highest doses tested. These findings demonstrated that the effect of (-)-linalool on pain responses is, at least partially, mediated by the activity of adenosine A1 and A2A receptors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D134
3,7-Dimethyl-1-propargylxanthine, ≥98% (HPLC), powder
C10H10N4O2