EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Glycoform-dependent conformational alteration of the Fc region of human immunoglobulin G1 as revealed by NMR spectroscopy.
PMID 16343775
Abstract
The Fc portion of immunoglobulin G (IgG) expresses the biantennary complex type oligosaccharides at Asn297 of the C(H)2 domain of each heavy chain with microheterogeneities depending on physiological and pathological states. These N-glycans are known to be essential for promotion of proper effector functions of IgG such as complement activation and Fcgamma receptor (FcgammaR)-mediated activities. To gain a better understanding of the role of Fc glycosylation, we prepared a series of truncated glycoforms of human IgG1-Fc and analyzed their interactions with human soluble FcgammaRIIIa (sFcgammaRIIIa) and with staphylococcal protein A by surface plasmon resonance and nuclear magnetic resonance (NMR) methods. Progressive but less pronounced reductions in the affinity for sFcgammaRIIIa were observed as a result of the galactosidase and subsequent N-acetylhexosaminidase treatments of IgG1-Fc. The following endoglycosidase D treatment, giving rise to a disaccharide structure composed of a fucosylated GlcNAc, abrogated the affinity of IgG1-Fc for sFcgammaRIIIa. On the other hand, those glycosidase treatments did not significantly affect the affinity of IgG1-Fc for protein A. Inspection of stable-isotope-assisted NMR data of a series of Fc glycoforms indicates that the stepwise trimming out of the carbohydrate residues results in concomitant increase in the number of amino acid residues perturbed thereby in the C(H)2 domains. Furthermore, the cleavage at the GlcNAcbeta1-4GlcNAc glycosidic linkage induced the conformational alterations of part of the lower hinge region, which makes no direct contact with the carbohydrate moieties and forms the major FcgammaR-binding site, while the conformation of the C(H)2/C(H)3 interface was barely perturbed that is the protein A-binding site. These results indicate that the carbohydrate moieties are required for maintaining the structural integrity of the FcgammaR-binding site.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
487910 Algal amino acid mixture-13C,15N, 98 atom % 13C, 98 atom % 15N