EMAIL THIS PAGE TO A FRIEND
Developmental and comparative immunology
Effects of linear cationic alpha-helical antimicrobial peptides on immune-relevant genes in trout macrophages.
PMID 16352337
Abstract
There are increasing evidence of the potential role of antimicrobial peptides in the regulation of immune responses in mammalian species. However, the effects of these peptides in fish have yet to be investigated. In this study, we examined the transcriptional expression profile of representative immune-relevant genes in a trout macrophage cell line, RTS11, in response to three linear cationic alpha-helical antimicrobial peptides (insect cecropin B, fish pleurocidin and a cecropin analogue CF17). The expression levels of two pro-inflammatory genes, interleukin-1 beta (IL-1 beta) and cyclo-oxygenase-2 (COX-2), increased in the peptide-treated RTS11 cells. The peptides did not appear to affect the expression levels of representative genes associated with antigen presentation, interferon response or JAK/STAT signal transduction. Furthermore, the induction of IL-1 beta and COX-2 in RTS11 by lipopolysaccharide was not adversely affected by these three antimicrobial peptides. Overall, the data indicate a pro-inflammatory effect of the three cationic antimicrobial peptides in the inflammatory response of salmonid species, suggesting a potential application of these peptides as immune adjuvant for fish vaccination.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C1796 Cecropin B, ≥97% (HPLC), powder
C176H301N51O42S