EMAIL THIS PAGE TO A FRIEND
Physical chemistry chemical physics : PCCP
Rotationally resolved electronic spectroscopy of tryptophol in the gas phase.
PMID 16358014
Abstract
High resolution S1-S0 fluorescence excitation spectra of tryptophol have been observed in the collision-free environment of a supersonic beam. Each origin band has been assigned to a unique conformer based on its observed rotational constants. Unlike its close relative tryptamine, which exhibits seven distinguishable conformers under similar conditions, tryptophol exhibits only four (GPy-in, GPh-in, and two anti structures). Possible reasons for this difference in behavior are discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T90301
3-(2-Hydroxyethyl)indole, 97%
C10H11NO