EMAIL THIS PAGE TO A FRIEND
Se pu = Chinese journal of chromatography
[Determination of acetochlor and oxyfluorfen by capillary gas chromatography].
PMID 16358708
Abstract
A method is described for the determination of acetochlor and oxyfluorfen by capillary gas chromatography with FID and an SE-30 capillary column (60 m x 0.53 mm i. d., 1.5 microm), using dibutyl phthalate as the internal standard. The standard deviations for acetochlor and oxyfluorfen concentration(mass fraction) were 0.44% and 0.47% respectively. The relative standard deviations for acetochlor and oxyfluorfen were 0.79% and 0.88% and the average recoveries for acetochlor and oxyfluorfen were 99.3% and 101.1% respectively. The method is simple, rapid and accurate.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
35031
Oxyfluorfen, PESTANAL®, analytical standard
C15H11ClF3NO4