EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
Enzymatic preparation of maltohexaose, maltoheptaose, and maltooctaose by the preferential cyclomaltooligosaccharide (cyclodextrin) ring-opening reaction of Pyrococcus furiosus thermostable amylase.
PMID 16364270
Abstract
Specific-length maltooligosaccharides, particularly maltohexaose, maltoheptaose, and maltooctaose, were prepared from cyclomaltooligosaccharides (cyclodextrins, CDs) by the preferential cyclodextrin ring-opening reaction of an amylolytic enzyme from Pyrococcus furiosus. The enzyme primarily produces maltohexaose, maltoheptaose, and maltooctaose by hydrolyzing alpha-, beta-, and gamma-CD, respectively. This study aims to develop a high-efficiency synthesis of specific maltooligosaccharides at high-purity. [formula: see text]
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M9153
Maltohexaose, ≥65% (HPLC)
C36H62O31