EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Optimization of a privileged structure leading to potent and selective human melanocortin subtype-4 receptor ligands.
PMID 16364639
Abstract
Design and synthesis of potent MC4 selective agonists based on cyclohexylpiperidine derived cyclic urea, oxazolidinones, and sulfonamide based privileged structures are disclosed.