EMAIL THIS PAGE TO A FRIEND
Journal of fluorescence
Synthesis and characterization of new fluorene-based singlet oxygen sensitizers.
PMID 16365698
Abstract
The synthesis, photophysical characterization, and determination of singlet oxygen quantum yields (Phi(Delta)) for a class of fluorene derivatives with potential application in two-photon photodynamic therapy (PDT) is reported. It has been demonstrated that these compounds possess the ability to generate singlet oxygen (1O2) upon excitation. A photochemical method, using 1,3-diphenylisobenzofuran (DPBF) as 1O2 chemical quencher, was employed to determine the singlet oxygen quantum yields (Phi(Delta)) of the fluorene-based photosensitizers in ethanol. Phi(Delta) values ranged from 0.35 to 0.75. These derivatives may have potential application as two-photon photosensitizers when pumped via two-photon excitation in the near-IR spectral region.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
105481
1,3-Diphenylisobenzofuran, 97%
C20H14O