EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Design, synthesis, and evaluation of pharmacological properties of cinnamic derivatives as antiatherogenic agents.
PMID 16366593
Abstract
A series of cinnamic and phosphonocinnamic derivatives have been synthesized and their ability to inhibit cell-mediated LDL oxidation and oxidized LDL-induced cytotoxicity was investigated. Electron-donating substituents surrounding the necessary 4-OH group of the aromatic ring showed the best results. Among the different series tested, amide 1, thioester 5c, phosphonoester 7c, and the fluorophosphonocinnamic analogue 12c exhibited a potent inhibitory effect against LDL oxidation (and subsequent toxicity) mediated by cultured human microvascular endothelial cells (HMEC-1), with an efficacy comparable to that observed with probucol. Beside this indirect protective effect, these compounds exhibited a direct protective effect against the toxicity of previously oxidized LDL in HMEC-1. These data suggest that the newly synthesized cinnamic compounds should protect against early events (cell-mediated LDL oxidation) occurring within the vascular wall in atherosclerosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P9672
Probucol, analytical standard
C31H48O2S2