EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Synthesis and biological properties of new alpha-methylene-gamma-butyrolactones and alpha,beta-unsaturated delta-lactones.
PMID 16368164
Abstract
The synthesis of a series of alpha-methylene-gamma-butyrolactones (compounds 4a, 4b, 6-12, 16, 17) and alpha,beta-unsaturated-delta-lactones (compounds 19-23, 25, 26) starting from 4,4-dimethyldihydrofuran-2,3-dione (1) has been described. Their chemical structures were assigned by spectroscopic evidence. These new compounds exhibited significantly different antiproliferative properties against cultured human tumor cell lines with their IC(50) values ranging from 0.88 to>20.00 microM.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
226416
α-Methylene-γ-butyrolactone, 97%
C5H6O2