EMAIL THIS PAGE TO A FRIEND
The Journal of Biological Chemistry
Role of the serum response factor in regulating contractile apparatus gene expression and sarcomeric integrity in cardiomyocytes.
PMID 16368687