EMAIL THIS PAGE TO A FRIEND
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Insulin stimulates L-carnitine accumulation in human skeletal muscle.
PMID 16368715
Abstract
Increasing skeletal muscle carnitine content may alleviate the decline in muscle fat oxidation seen during intense exercise. Studies to date, however, have failed to increase muscle carnitine content, in healthy humans, by dietary or intravenous L-carnitine administration. We hypothesized that insulin could augment Na+-dependent skeletal muscle carnitine transport. On two randomized visits, eight healthy men underwent 5 h of intravenous L-carnitine infusion with serum insulin maintained at fasting (7.4+/-0.4 mIU*l(-1)) or physiologically high (149.2+/-6.9 mIU*l(-1)) concentrations. The combination of hypercarnitinemia (approximately 500 micromol*l(-1)) and hyperinsulinemia increased muscle total carnitine (TC) content from 22.0 +/- 0.9 to 24.7 +/- 1.4 mmol*(kg dm)(-1) (P<0.05) and was associated with a 2.3 +/- 0.3-fold increase in carnitine transporter protein (OCTN2) mRNA expression (P<0.05). Hypercarnitinemia in the presence of a fasting insulin concentration had no effect on either of these parameters. This study demonstrates that insulin can acutely increase muscle TC content in humans during hypercarnitinemia, which is associated with an increase in OCTN2 transcription. These novel findings may be of importance to the regulation of muscle fat oxidation during exercise, particularly in obesity and type 2 diabetes where it is known to be impaired.