EMAIL THIS PAGE TO A FRIEND
Oncogene
Protocadherin 15 (PCDH15): a new secreted isoform and a potential marker for NK/T cell lymphomas.
PMID 16369489
Abstract
Natural killer cells are well known to play an important role in immune defense against tumor development and viral infections. To further characterize new functionally relevant structures in these cells, we studied a series of monoclonal antibodies that we have raised against the NK cell line YT. One of these antibodies previously described as AY19, recognizes a 85 kD surface glycoprotein. Here we report the identification of a new secreted isoform of protocadherin 15, PCDH15C, which represents a potential associated protein for p85. Importantly, whereas protocadherins are absent from the surface of normal hematopoietic cells, we describe, for the first time, that PCDH15 is expressed in cytotoxic tumor-derived T- and NK-cell lines as well as in biopsies of nasal NK/T-cell lymphomas.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV50153
Anti-PCDH15 antibody produced in rabbit, affinity isolated antibody