EMAIL THIS PAGE TO A FRIEND
Genes to cells : devoted to molecular & cellular mechanisms
Evaluation of somatic alterations of Pcdh-alpha transcripts in the brain by cDNA analysis without PCR.
PMID 16371135