EMAIL THIS PAGE TO A FRIEND
Phytotherapy research : PTR
Antibacterial activity of polygodial.
PMID 16372365
Abstract
Polygodial was found to possess moderate antibacterial activity against gram-positive bacteria including Bacillus subtilis, Staphylococcus aureus and gram-negative bacteria including Escherichia coli and Salmonella choleraesuis with minimum bactericidal concentrations (MBC) of 100 and 100 microg/mL (0.34 mm) and 100 and 50 microg/mL, respectively. The time kill curve study showed that polygodial was bactericidal against B. subtilis and S. choleraesuis. However, there was a difference in its bactericidal action against endospore-forming B. subtilis and food-borne S. choleraesuis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0049 Polygodial, ≥97% (HPLC)
C15H22O2