EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Ascorbic acid reduction of microtubule protein disulfides and its relevance to protein S-nitrosylation assays.
PMID 16375859
Abstract
The biotin switch assay was developed to aid in the identification of S-nitrosylated proteins in different cell types. However, our work with microtubule proteins including tubulin and its associated proteins tau and microtubule-associated protein-2 shows that ascorbic acid is not a selective reductant of protein S-nitrosothiols as described in the biotin switch assay. Herein we show that ascorbic acid reduces protein disulfides in tubulin, tau, and microtubule-associated protein-2 that are formed by peroxynitrite anion. Reduction of microtubule-associated protein disulfides by ascorbic acid following peroxynitrite treatment restores microtubule polymerization kinetics to control levels. We also show that ascorbic acid reduces the disulfide dithiobis(2-nitrobenzoic acid), a reagent commonly used to detect protein thiols. Not only do we describe a new reactivity of ascorbic acid with microtubule proteins but we expose an important limitation when using the biotin switch assay to detect protein S-nitrosylation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I9271
5-(Iodoacetamido)fluorescein, ≥90% (HPLC)
C22H14INO6