EMAIL THIS PAGE TO A FRIEND
Bioscience, biotechnology, and biochemistry
Isolation and functional analysis of cytochrome P450 CYP153A genes from various environments.
PMID 16377903
Abstract
The cytochrome P450 CYP153 family is thought to mediate the terminal hydroxylation reactions of n-alkanes. We isolated 16 new P450 CYP153A genes (central region) from various environments such as petroleum-contaminated soil and groundwater, as well as one from the n-alkane-degrading bacterium Alcanivorax borkumensis SK2 (designated P450balk). The sequences of the new P450 genes were extended by PCR to generate full-length chimeric P450 genes, using the N- and C-terminal domains of P450balk. A differential CO-reduced P450 spectral analysis indicated that 8 P450 genes among the 16 chimeric genes were expressed in Escherichia coli to generate a soluble and functional enzyme. The several functional chimeric P450s and P450balk were further fused to the reductase domain of the self-sufficient P450 monooxygenase (P450RhF) at the C-terminus. E. coli cells expressing these self-sufficient P450 chimeric genes converted n-alkanes, cyclohexane, 1-octene, n-butylbenzene, and 4-phenyl-1-butene into 1-alkanols, cyclohexanol, 1,2-epoxyoctane, 1-phenyl-4-butanol, and 2-phenethyl-oxirane, respectively.