EMAIL THIS PAGE TO A FRIEND
Biomedical chromatography : BMC
Peroxyoxalate chemiluminescence detection of condensates of malondialdehyde with thiobarbituric acids using a flow system.
PMID 1638090
Abstract
The peroxyoxalate chemiluminescence(CL) detection method for the evaluation of the CL intensity of malondialdehyde(MDA) condensates with seven 2-thiobarbituric acid derivatives is described. The method consists of a flow injection technique together with a CL detection system using bis(2,4,6-trichlorophenyl) oxalate(TCPO) and hydrogen peroxide as chemiluminogenic reagents. Linear correlations between CL intensity and concentration are obtained for pmol levels of condensates. Among the condensates, 1,3-diethyl-2-thiobarbituric acid(DETBA)-MDA shows the largest CL intensity. High performance liquid chromatography (HPLC)/CL detection of DETBA-MDA and 1,3-diphenyl-2-thiobarbituric acid(DPTBA)-MDA using a mixture of TCPO and hydrogen peroxide in acetonitrile as a postcolumn reagent solution is also described. The detection limits for DETBA-MDA and DPTBA-MDA are 20 and 200 fmol, respectively, per 20 microL injection at a signal-to-noise ratio of 2. This HPLC/CL detection system was applied to the determination of MDA in rat brains by using DETBA as a fluorescent derivatizing reagent.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
161950
1,3-Diethyl-2-thiobarbituric acid, 99%
C8H12N2O2S