EMAIL THIS PAGE TO A FRIEND
Molecular and cellular biology
Inhibitor of apoptosis protein cIAP2 is essential for lipopolysaccharide-induced macrophage survival.
PMID 16382159
Abstract
The cellular inhibitor of apoptosis 2 (cIAP2/HIAP1) is a potent inhibitor of apoptotic death. In contrast to the other members of the IAP family, cIAP2 is transcriptionally inducible by nuclear factor-kappaB in response to multiple triggers. We demonstrate here that cIAP2-/- mice exhibit profound resistance to lipopolysaccharide (LPS)-induced sepsis, specifically because of an attenuated inflammatory response. We show that LPS potently upregulates cIAP2 in macrophages and that cIAP2-/- macrophages are highly susceptible to apoptosis in a LPS-induced proinflammatory environment. Hence, cIAP2 is critical in the maintenance of a normal innate immune inflammatory response.