EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Synthesis of chalcone analogues with increased antileishmanial activity.
PMID 16386424
Abstract
Eighteen analogues of an active natural chalcone were synthesized using xanthoxyline and some derivatives, and these analogues were tested for selective activity against both promastigotes and intracellular amastigotes of Leishmania amazonensis in vitro. Three analogues (10, 12, and 19) containing nitro, fluorine or bromine groups, respectively, displayed increased selective activity against the parasites as compared with the natural chalcone. The nitrosylated chalcone 10 was also tested intralesionally in infected mice and was found to be as effective as Pentostan reference drug at a dose 100 times higher than that of the chalcone in controlling both the lesion growth and the parasite burden.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
630586
2′-Hydroxy-4′,6′-dimethoxyacetophenone, 97%
C10H12O4