EMAIL THIS PAGE TO A FRIEND
International journal of hygiene and environmental health
Sodium dichloroisocyanurate (NaDCC) tablets as an alternative to sodium hypochlorite for the routine treatment of drinking water at the household level.
PMID 16387550
Abstract
Household water treatment using sodium hypochlorite (NaOCl) has been recognized as a cost-effective means of reducing the heavy burden of diarrhea and other waterborne diseases, especially among populations without access to improved water supplies. Sodium dichloroisocyanurate (NaDCC), which is widely used in emergencies, is an alternative source of chlorine that may present certain advantages over NaOCl for household-based interventions in development settings. We summarize the basic chemistry and possible benefits of NaDCC, and review the available literature concerning its safety and regulatory treatment and microbiological effectiveness. We review the evidence concerning NaDCC in field studies, including microbiological performance and health outcomes. Finally, we examine studies and data to compare NaDCC with NaOCl in terms of compliance, acceptability, affordability and sustainability, and suggest areas for further research.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
218928
Sodium dichloroisocyanurate, 96%
C3Cl2N3NaO3
35915
Sodium dichloroisocyanurate dihydrate, ≥98.0% (AT)
C3Cl2N3NaO3 · 2H2O