EMAIL THIS PAGE TO A FRIEND
Applied and environmental microbiology
Expression of the ubiE gene of Geobacillus stearothermophilus V in Escherichia coli K-12 mediates the evolution of selenium compounds into the headspace of selenite- and selenate-amended cultures.
PMID 16391146
Abstract
The ubiE gene of Geobacillus stearothermophilus V, with its own promoter, was cloned and introduced into Escherichia coli. The cloned gene complemented the ubiE gene deficiency of E. coli AN70. In addition, the expression of this gene in E. coli JM109 resulted in the evolution of volatile selenium compounds when these cells were grown in selenite- or selenate-amended media. These compounds were dimethyl selenide and dimethyl diselenide.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
328502
Dimethyl diselenide, 96%
C2H6Se2
41572
Dimethyl selenide, ≥99.0% (GC)
C2H6Se