EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Enantioselective total syntheses of (-)-clasto-lactacystin beta-lactone and 7-epi-(-)-clasto-lactacystin beta-lactone.
PMID 16391758
Abstract
An alkylidene carbene 1,5-CH insertion has been used as a key step in an efficient enantioselective total synthesis of (-)-clasto-lactacystin beta-lactone, and its C7-epimer. An additional noteworthy feature of the synthesis is the use of a novel oxidative deprotection procedure, utilizing DMDO, for the conversion of a late-stage benzylidene acetal into a primary alcohol and a secondary benzoate ester.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L7035
clasto-Lactacystin β-lactone
C10H15NO4